missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-OXPAT, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of the mouse OXPAT protein
Antigen OXPAT
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11060511 Novus Biologicals Canada
NB11060511
0.1mL Each for $329.00
NB11060511T Novus Biologicals Canada
NB11060511SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
OXPAT
Affinity Purified
RUO
Human and mouse. Immunogen sequence has 89% homology to rat and 75% homology to bovine.
PBS, 30% glycerol
A synthetic peptide made to a C-terminal region of the mouse OXPAT protein
Polyclonal
Primary
Simple Western,Western Blot
440503
1.01mg/mL