Please login to your online account to display your discounted pricing

anti-OXPAT, Polyclonal, Novus Biologicals™

$144.63 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region of the human OXPAT protein
Antigen OXPAT
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11060509T

 
novus biologicals canada
NB11060509SS
Unlabeled 0.025mL Each for $144.63

NB11060509

 
novus biologicals canada
NB11060509
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a region of the human OXPAT protein
Antigen OXPAT
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Test Specificity Human, bovine and mouse. Immunogen has 94% homology to sheep and porcine and 84% homology to rat.
Gene Accession No. Q00G26
Gene ID (Entrez) 440503
Formulation PBS, 30% glycerol
Concentration 1.0mg/mL