Please login to your online account to display your discounted pricing

anti-Otx2, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 10-60 of human OTX2 was used as the immunogen.
Antigen Otx2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224652SS

 
novus biologicals canada
NBP224652SS
Unlabeled 0.025mg Each for $109.00

NBP224652

 
novus biologicals canada
NBP224652
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 10-60 of human OTX2 was used as the immunogen.
Antigen Otx2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, wild boar, horse, mouse, chimpanzee, rhesus monkey, dog and sheep, 94% homologous in opossum and 77% homologous in chicken and Xenopus.
Gene ID (Entrez) 5015
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL