Please login to your online account to display your discounted pricing

anti-Otx2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224652SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224652SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.05% BSA
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, wild boar, horse, mouse, chimpanzee, rhesus monkey, dog and sheep, 94% homologous in opossum and 77% homologous in chicken and Xenopus.
Conjugate Unlabeled
Immunogen A portion of amino acids 10-60 of human OTX2 was used as the immunogen.
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Host Species Rabbit
Antigen Otx2
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5015
Quantity 0.025mg
Regulatory Status RUO
Format Purified
Applications Western Blotting