Please login to your online account to display your discounted pricing

anti-Otx2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224652

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224652

  • / Each
 

Description & Specifications

Specifications

Antigen Otx2
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 10-60 of human OTX2 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5015
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, wild boar, horse, mouse, chimpanzee, rhesus monkey, dog and sheep, 94% homologous in opossum and 77% homologous in chicken and Xenopus.
Target Species Canine
Target Species Chicken
Target Species Equine
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat