Please login to your online account to display your discounted pricing

anti-OS9, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of isoform 1 of the human OS9 protein (within residues 300-400). [Swiss-Prot No. Q13438]
Antigen OS9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100519

 
novus biologicals canada
NB100519
Unlabeled 0.1mL Each for $309.05

NB100519SS

 
novus biologicals canada
NB100519SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of isoform 1 of the human OS9 protein (within residues 300-400). [Swiss-Prot No. Q13438]
Antigen OS9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This is specific for isoforms 1 and 2 of human OS-9.
Gene Accession No. Q13438
Gene ID (Entrez) 10956
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL