Please login to your online account to display your discounted pricing

anti-OS9, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen 2
Antigen OS9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

BC100520

 
novus biologicals canada
BC100520
Unlabeled 0.1mL Each for $309.05

BC100520SS

 
novus biologicals canada
BC100520SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen 2
Antigen OS9
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Test Specificity Human, mouse and rat (see customer review).
Gene Accession No. Q13438
Gene ID (Entrez) 10956
Formulation Tris-glycine, 150mM NaCl
Concentration 0.2mg/mL