Please login to your online account to display your discounted pricing

anti-OLFM4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224535

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224535

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 10562
Gene Accession No. Q6UX06
Conjugate Unlabeled
Primary or Secondary Primary
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Immunogen A synthetic peptide corresponding to amino acids between 400 and 450 of human OLFM4 was used as immunogen for this antibody.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoform CRA_a) and chimpanzee, 85% in monkey, mouse and rat, 76% in cow and 75% homologous in dog.
Antigen OLFM4
Host Species Rabbit
Quantity 0.1mg
Regulatory Status RUO
Format Affinity Purified