Please login to your online account to display your discounted pricing

anti-OLFM4, Polyclonal, Novus Biologicals™

$109.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids between 400 and 450 of human OLFM4 was used as immunogen for this antibody.
Antigen OLFM4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224535

 
novus biologicals canada
NBP224535
Unlabeled 0.1mg Each for $362.00

NBP224535SS

 
novus biologicals canada
NBP224535SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids between 400 and 450 of human OLFM4 was used as immunogen for this antibody.
Antigen OLFM4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoform CRA_a) and chimpanzee, 85% in monkey, mouse and rat, 76% in cow and 75% homologous in dog.
Gene Accession No. Q6UX06
Gene ID (Entrez) 10562
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL