Please login to your online account to display your discounted pricing

anti-OGG1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100106

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100106

  • / Each
 

Description & Specifications

Specifications

Antigen OGG1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. O15527
Host Species Rabbit
Immunogen A peptide derived from human Ogg1 (within amino acids 1-100). [UniProt No. O15527]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4968
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat