Please login to your online account to display your discounted pricing

anti-OCT6, Schwann Cell Marker, Polyclonal, Novus Biologicals™

$363.90 - $363.90

Specifications

Host Species Rabbit
Immunogen Synthetic peptide
Antigen OCT6XXXXX Schwann Cell Marker
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP119586

 
novus biologicals canada
NBP119586
Unlabeled 0.1mg Each for $363.90
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide
Antigen OCT6XXXXX Schwann Cell Marker
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity Oct-6 (A332) detects endogenous levels of Oct-6 protein.
Gene Accession No. Q03052
Gene ID (Entrez) 5453
Formulation phosphate buffered saline (PBS), approx. pH 7.2.
Concentration 1.0mg/mL