Please login to your online account to display your discounted pricing

anti-OCT6, Schwann Cell Marker, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP119586SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP119586SS

  • / Each
 

Description & Specifications

Specifications

Formulation phosphate buffered saline (PBS), approx. pH 7.2.
Gene ID (Entrez) 5453
Concentration 1.0mg/mL
Quantity 0.025mg
Conjugate Unlabeled
Gene Accession No. Q03052
Monoclonal or Polyclonal Polyclonal
Immunogen Synthetic peptide
Primary or Secondary Primary
Test Specificity Oct-6 (A332) detects endogenous levels of Oct-6 protein.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen OCT6XXXXX Schwann Cell Marker
Format Affinity Purified
Applications Western Blotting
Regulatory Status RUO
Host Species Rabbit