Please login to your online account to display your discounted pricing

anti-OCT6, Schwann Cell Marker, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP119586

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP119586

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation phosphate buffered saline (PBS), approx. pH 7.2.
Conjugate Unlabeled
Gene ID (Entrez) 5453
Test Specificity Oct-6 (A332) detects endogenous levels of Oct-6 protein.
Gene Accession No. Q03052
Concentration 1.0mg/mL
Immunogen Synthetic peptide
Regulatory Status RUO
Format Affinity Purified
Primary or Secondary Primary
Applications Western Blotting
Antigen OCT6XXXXX Schwann Cell Marker
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Rabbit
Quantity 0.1mg