Please login to your online account to display your discounted pricing

anti-OCT4, Embryonic Stem Cell Marker, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002379

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002379

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 5460
Primary or Secondary Primary
Test Specificity Human, monkey, rat, mouse. Immunogen has 93% homology to cow, feline and pig. Customer feedback suggests that this antibody does not give good results in IHC-P on feline tissues.
Formulation Tris-glycine, 150mM NaCl
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Immunogen A synthetic peptide made to an internal portion of the human OCT4 protein (between residues 100-200) [UniProt No. Q01860]
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene Accession No. Q01860
Concentration 0.85mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Regulatory Status RUO
Antigen OCT4XXXXX Embryonic Stem Cell Marker
Host Species Rabbit
Quantity 0.1mL
Format Affinity Purified