Please login to your online account to display your discounted pricing

anti-OCT4 Embryonic Stem Cell Marker, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224665SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224665SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 5460
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Antigen OCT4 Embryonic Stem Cell Marker
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a,) chimpanzee, and monkey, 92% homologous in cow and dog, 85% homologous in horse, and rat and 78% homologous in mouse.
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Regulatory Status RUO
Immunogen Within the range of amino acids 230-257 of human OCT4 protein was used as the immunogen.
Host Species Rabbit
Quantity 0.025mg
Format Purified