Please login to your online account to display your discounted pricing

anti-OCT4 Embryonic Stem Cell Marker, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 230-257 of human OCT4 protein was used as the immunogen.
Antigen OCT4 Embryonic Stem Cell Marker
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224665SS

 
novus biologicals canada
NBP224665SS
Unlabeled 0.025mg Each for $109.00

NBP224665

 
novus biologicals canada
NBP224665
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 230-257 of human OCT4 protein was used as the immunogen.
Antigen OCT4 Embryonic Stem Cell Marker
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a,) chimpanzee, and monkey, 92% homologous in cow and dog, 85% homologous in horse, and rat and 78% homologous in mouse.
Gene ID (Entrez) 5460
Formulation 100μg in 200μL PBS containing 0.05% BSA