Please login to your online account to display your discounted pricing

anti-OCT1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human OCT-1 was used as the immunogen.
Antigen OCT1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224677

 
novus biologicals canada
NBP224677
Unlabeled 0.1mg Each for $257.41

NBP224677SS

 
novus biologicals canada
NBP224677SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human OCT-1 was used as the immunogen.
Antigen OCT1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, monkey, 83% homologous in mouse, 77% homologous in cow and 72% homologous in dog and rat.
Gene ID (Entrez) 5451
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL