Please login to your online account to display your discounted pricing

anti-OCT1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 410-460 of human OCT1/OTF1 was used as the immunogen.
Antigen OCT1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224499

 
novus biologicals canada
NBP224499
Unlabeled 0.1mg Each for $257.41

NBP224499SS

 
novus biologicals canada
NBP224499SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 410-460 of human OCT1/OTF1 was used as the immunogen.
Antigen OCT1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, macaque, chimpanzee, rhesus monkey, and dog, 94% homologous in horse, and 88% homologous in rat, mouse and opossum.
Gene ID (Entrez) 5451
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL