Please login to your online account to display your discounted pricing

anti-OCT1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224499

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224499

  • / Each
 

Description & Specifications

Specifications

Antigen OCT1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 410-460 of human OCT1/OTF1 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 5451
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, macaque, chimpanzee, rhesus monkey, and dog, 94% homologous in horse, and 88% homologous in rat, mouse and opossum.
Target Species Bovine
Target Species Equine
Target Species Human
Target Species Murine
Target Species Rat