missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Nup53, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 50-100 of human NUP35 was used as the immunogen for the antibody.
Antigen Nup53
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224525 Novus Biologicals Canada
NBP224525
0.1mg Each for $257.41
NBP224525SS Novus Biologicals Canada
NBP224525SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Nup53
Affinity Purified
RUO
Human reactivity. Predicted in mouse, rat, bovine, canine, equine, most primate species (93% or better), and rabbit based on homology.
129401
0.5mg/mL
A portion of amino acids 50-100 of human NUP35 was used as the immunogen for the antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
Q8NFH5
100μg antibody in 200μL PBS containing 0.05% BSA