Please login to your online account to display your discounted pricing

anti-Nup53, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224525SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224525SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 129401
Conjugate Unlabeled
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q8NFH5
Concentration 0.5mg/mL
Format Affinity Purified
Cross Reactivity Human
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Test Specificity Human reactivity. Predicted in mouse, rat, bovine, canine, equine, most primate species (93% or better), and rabbit based on homology.
Host Species Rabbit
Regulatory Status RUO
Antigen Nup53
Immunogen A portion of amino acids 50-100 of human NUP35 was used as the immunogen for the antibody.
Quantity 0.025mg