Please login to your online account to display your discounted pricing

anti-Nup53, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224525

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224525

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene ID (Entrez) 129401
Test Specificity Human reactivity. Predicted in mouse, rat, bovine, canine, equine, most primate species (93% or better), and rabbit based on homology.
Immunogen A portion of amino acids 50-100 of human NUP35 was used as the immunogen for the antibody.
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Rabbit
Format Affinity Purified
Concentration 0.5mg/mL
Gene Accession No. Q8NFH5
Primary or Secondary Primary
Quantity 0.1mg
Antigen Nup53
Regulatory Status RUO
Cross Reactivity Human