missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
NUMB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen PA116831
Description
The target sequence has 90% sequence homology with rat.
NUMB Polyclonal antibody specifically detects NUMB in Chicken, Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
NUMB | |
Polyclonal | |
Unconjugated | |
Numb | |
c14_5527; C14orf41; h-Numb; mnb; m-Nb; m-numb; Nb; NUMB; NUMB endocytic adaptor protein; numb gene homolog; numb gene homolog (Drosophila); numb homolog; numb homolog (Drosophila); NUMB isoform 3; Numb variant 1; Numb variant 2; NUMB, endocytic adaptor protein; protein numb homolog; Protein S171; PTB domain protein; S171 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18222, 395639, 8650 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
3.5 mg/mL | |
tris glycine with 0.15M NaCl and 0.05% sodium azide | |
P49757, Q9QZS3 | |
Numb | |
A synthetic peptide made to a C-terminal region of the murine numb protein sequence (between residues 600-653). | |
100 μL | |
Primary | |
Chicken, Human, Mouse | |
Antibody | |
IgG |