missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Numb, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 12-40 of human NUMB was used as the immunogen for the antibody
Antigen Numb
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227181 Novus Biologicals Canada
NBP227181
0.1mg Each for $255.80
NBP227181SS Novus Biologicals Canada
NBP227181SS
0.025mg Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
Numb
Affinity Purified
RUO
The amino acid sequence used as immunogen for the NUMB antibody is 100% homologous in human, mouse, rat, cow and monkey.
8650
0.5mg/mL
A synthetic peptide corresponding to amino acids 12-40 of human NUMB was used as the immunogen for the antibody
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
P49757
100μg in 200μL PBS containing 0.05% BSA.