Please login to your online account to display your discounted pricing

anti-Nucleolin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600241SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600241SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4691
Conjugate Unlabeled
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Plant
Cross Reactivity Rat
Applications Chromatin Immunoprecipitation
Applications Electron Microscopy
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunomicroscopy
Applications Simple Western
Applications Western Blotting
Host Species Rabbit
Gene Accession No. P19338
Test Specificity Human, mouse, rat and soy bean plant.
Regulatory Status RUO
Antigen Nucleolin
Quantity 0.025mL
Immunogen A fusion protein containing amino acids 284-709 of human Nucleolin. [UniProt No. P19338]
Format Antiserum