Please login to your online account to display your discounted pricing

anti-Neurotrophin-3 Polyclonal, Thermo Scientific™

Neurotrophin-3 Polyclonal Ab PA1-18384 from Thermo Scientific™ detects Neurotrophin-3 in Ck, Hu and Rt samples, and is validated for ELISA, IHC, Inhibition Assays and WB

$543.75 - $543.75

Specifications

Antigen Neurotrophin-3
Cross Reactivity Human
Cross Reactivity Non-human primate
Cross Reactivity Rat
Format Purified
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-636

 
pierce biotechnology inc
PA118384
Unlabeled 50μg Each for $543.75

01-670-641

 
pierce biotechnology inc
PA118389
Unlabeled 500μg Each for $543.75

01-670-643

 
pierce biotechnology inc
PA118391
Unlabeled 500μg Each for $543.75
Description & Specifications

Specifications

Antigen Neurotrophin-3
Cross Reactivity Human
Cross Reactivity Non-human primate
Cross Reactivity Rat
Format Purified
Host Species Rabbit
Immunogen Recombinant human NT3.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Neurotrophin-3 Polyclonal Ab PA1-18384 from Thermo Scientific™ detects Neurotrophin-3 in Ck, Hu and Rt samples, and is validated for ELISA, IHC, Inhibition Assays and WB

The protein encoded by this gene is a member of the neurotrophin family, that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.
Host Species: Sheep
Cross Reactivity: Chicken, Human, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues Y(139) A E H K S H R G E Y(149) of human, mouse and rat NT3.
Formulation: 50μg purified Antibody (1mg/mL) no preservative;
Synonyms: Neurotrophin-3, Neurotrophic factor, HDNF, Nerve growth factor 2, NGF-2, Ntf3, Ntf-3
Uniprot Accession: P18280, P20783, P25433
Gene Symbol: NTF3
Entrez GeneID: 428099, 4908, 81737
PA1-18384 detects Neurotrophin-3 in Chicken, Human and Rat samples. PA1-18384 has been successfully used in ELISA, Immunohistochemistry, Inhibition Assays and Western Blot procedures. PA1-18384 The immunogen is Synthetic peptide corresponding to residues Y(139) A E H K S H R G E Y(149) of human, mouse and rat NT3..