Please login to your online account to display your discounted pricing

anti-NSD3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP104938

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP104938

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation Tris-citrate/phosphate, pH 7-8
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 200-300). [Swiss-Prot No. Q9BZ95]
Regulatory Status RUO
Cross Reactivity Human
Gene Accession No. Q9BZ95
Host Species Rabbit
Applications Western Blotting
Gene ID (Entrez) 54904
Test Specificity This has been observed to recognize a band that corresponds to isoform 2 of NSD3. Human. Predicted to react with mouse based on 100% sequence homology. Immunogen has 91% homology to bovine.
Concentration 1mg/mL
Conjugate Unlabeled
Antigen NSD3
Quantity 0.1mL