Please login to your online account to display your discounted pricing

anti-NSD3, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 500-600). [Swiss-Prot No. Q9BZ95]
Antigen NSD3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP104991

 
novus biologicals canada
NBP104991
Unlabeled 0.1mL Each for $255.80

NBP104991SS

 
novus biologicals canada
NBP104991SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 500-600). [Swiss-Prot No. Q9BZ95]
Antigen NSD3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This has been observed to recognize a band that corresponds to isoform 3 of NSD3. Human. Immunogen sequence has 90% homology with bovine.
Gene Accession No. Q9BZ95
Gene ID (Entrez) 54904
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL