Please login to your online account to display your discounted pricing

anti-NSD3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP104991SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP104991SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Test Specificity This has been observed to recognize a band that corresponds to isoform 3 of NSD3. Human. Immunogen sequence has 90% homology with bovine.
Conjugate Unlabeled
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q9BZ95
Applications Western Blotting
Format Affinity Purified
Host Species Rabbit
Concentration 1mg/mL
Cross Reactivity Human
Primary or Secondary Primary
Gene ID (Entrez) 54904
Immunogen Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 500-600). [Swiss-Prot No. Q9BZ95]
Quantity 0.025mL
Regulatory Status RUO
Antigen NSD3