Please login to your online account to display your discounted pricing

anti-NSD3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP104991

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP104991

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 54904
Test Specificity This has been observed to recognize a band that corresponds to isoform 3 of NSD3. Human. Immunogen sequence has 90% homology with bovine.
Primary or Secondary Primary
Conjugate Unlabeled
Formulation Tris-citrate/phosphate, pH 7-8
Host Species Rabbit
Quantity 0.1mL
Antigen NSD3
Format Affinity Purified
Monoclonal or Polyclonal Polyclonal
Gene Accession No. Q9BZ95
Immunogen Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 500-600). [Swiss-Prot No. Q9BZ95]
Concentration 1mg/mL
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Human