Please login to your online account to display your discounted pricing

anti-NSD3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP104942SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP104942SS

  • / Each
 

Description & Specifications

Specifications

Concentration 1mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. Q9BZ95
Host Species Rabbit
Cross Reactivity Human
Antigen NSD3
Test Specificity This has been observed to recognize a band that corresponds to isoform 1 of NSD3. Human. Predicted to react with mouse and bovine based on 100% sequence homology.
Formulation Tris-citrate/phosphate, pH 7-8
Gene ID (Entrez) 54904
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of the human NSD3 protein (within residues 300-400). [Swiss-Prot No. Q9BZ95]
Quantity 0.025mL
Applications Western Blotting