Please login to your online account to display your discounted pricing

anti-NS1-BP, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 24-84 of human Influenza A H1N1 nonstructural protein were used as the immunogen.
Antigen NS1-BP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224567

 
novus biologicals canada
NBP224567
Unlabeled 0.1mg Each for $309.05

NBP224567SS

 
novus biologicals canada
NBP224567SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 24-84 of human Influenza A H1N1 nonstructural protein were used as the immunogen.
Antigen NS1-BP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous inhuman, chimpanzee, mouse,94% homologous inrat,88% homologous inchicken, cow, opossum, monkey and 83% homologous in dog.
Gene ID (Entrez) 10625
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL