Please login to your online account to display your discounted pricing

anti-NR5A1 Polyclonal, Thermo Scientific™


NR5A1 Polyclonal antibody PA1-30599 from Thermo Scientific™ specifically detects NR5A1 in Human samples, and it is validated for Western Blot

Manufacturer: pierce biotechnology inc  PA130599

 View more versions of this product

Catalog No. PIPA130599

  • / Each
 

Description & Specifications

Specifications

Formulation 50μg purified antibody (1mg/mL) in PBS with 2% sucrose and no preservative
Gene ID (Entrez) 2516
Primary or Secondary Primary
Format Purified
Gene Accession No. Q13285
Gene Alias AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1
Quantity 50μg
Species Reactivity Human
Storage Requirements Avoid Freeze/Thaw Cycles
Monoclonal or Polyclonal Polyclonal
Antigen NR5A1
Host Species Rabbit
Immunogen Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human NR5A1
Cross Reactivity Human
Isotype IgG
Applications Western Blotting
Regulatory Status RUO
Conjugate Unlabeled
Classification Polyclonal
Gene Symbols NR5A1
Type Primary

NR5A1 Polyclonal antibody PA1-30599 from Thermo Scientific™ specifically detects NR5A1 in Human samples, and it is validated for Western Blot

The protein encoded by this gene is a transcriptional activator involved in sex determination. The encoded protein binds DNA as a monomer. Defects in this gene are a cause of XY sex reversal with or without adrenal failure as well as adrenocortical insufficiency without ovarian defect.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human NR5A1
Formulation: 50μg purified antibody (1mg/mL) in PBS with 2% sucrose and no preservative
Synonyms: AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1
Uniprot Accession: Q13285
Entrez GeneID: 2516
Gene Symbol: NR5A1
PA1-30599 detects NR5A1 from human samples. PA1-30599 has been successfully used in Western blot applications. The PA1-30599 immunogen is: Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human NR5A1