Please login to your online account to display your discounted pricing

anti-NR5A1 Polyclonal, Thermo Scientific™

NR5A1 Polyclonal antibody PA1-30599 from Thermo Scientific™ specifically detects NR5A1 in Human samples, and it is validated for Western Blot

$616.25 - $616.25

Specifications

Antigen NR5A1
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

PIPA130599

 
pierce biotechnology inc
PA130599
Each for $616.25
Description & Specifications

Specifications

Antigen NR5A1
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Host Species Rabbit
Quantity 50μg
Regulatory Status RUO
Type Primary
Formulation 50μg purified antibody (1mg/mL) in PBS with 2% sucrose and no preservative
Gene Symbols NR5A1
Immunogen Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human NR5A1
Isotype IgG
Storage Requirements Avoid Freeze/Thaw Cycles

NR5A1 Polyclonal antibody PA1-30599 from Thermo Scientific™ specifically detects NR5A1 in Human samples, and it is validated for Western Blot

The protein encoded by this gene is a transcriptional activator involved in sex determination. The encoded protein binds DNA as a monomer. Defects in this gene are a cause of XY sex reversal with or without adrenal failure as well as adrenocortical insufficiency without ovarian defect.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human NR5A1
Formulation: 50μg purified Antibody (1mg/mL) in PBS, with 2% sucrose and no preservative;
Synonyms: AD4BP, ELP, FTZ1, FTZF1, POF7, SF-1, SF1
Uniprot Accession: Q13285
Gene Symbol: NR5A1
Entrez GeneID: 2516
PA1-30599 detects NR5A1 in Human samples. PA1-30599 has been successfully used in Western Blot procedures. PA1-30599 The immunogen is Polyclonal antibody produced in rabbits immunized with a synthetic peptide corresponding to a region of human NR5A1.