Please login to your online account to display your discounted pricing

anti-NQO-1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 250-274 of human NQO1 was used as the immunogen.
Antigen NQO-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223792

 
novus biologicals canada
NBP223792
Unlabeled 0.1mL Each for $259.02

NBP223792SS

 
novus biologicals canada
NBP223792SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acid 250-274 of human NQO1 was used as the immunogen.
Antigen NQO-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey, chimpanzee and dog, 95% homologous in rat, 90% homologous in mouse and 83% homologous in chicken.
Gene Accession No. P15559
Gene ID (Entrez) 1728
Formulation 100μL of crude rabbit sera with 50% glycerol