Please login to your online account to display your discounted pricing

anti-NPY2R, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human NPY2-R was used as the immunogen.
Antigen NPY2R
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056480

 
novus biologicals canada
NB10056480
Unlabeled 0.1mg Each for $255.80

NB10056480T

 
novus biologicals canada
NB10056480SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human NPY2-R was used as the immunogen.
Antigen NPY2R
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Reacts with Human and Rhesus Monkey. 83% sequence identity with canine and mouse proteins.
Gene Accession No. NP_000901
Gene ID (Entrez) 4887
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL