Please login to your online account to display your discounted pricing

anti-NPY2R, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056480SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056480T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 4887
Test Specificity Reacts with Human and Rhesus Monkey. 83% sequence identity with canine and mouse proteins.
Antigen NPY2R
Host Species Rabbit
Quantity 0.025mg
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Formulation PBS containing 0.2% gelatin
Gene Accession No. NP_000901
Immunogen A portion of amino acids 350-400 of human NPY2-R was used as the immunogen.
Format Affinity Purified
Cross Reactivity Human
Cross Reactivity Primate
Regulatory Status RUO