Please login to your online account to display your discounted pricing

anti-NPY2R, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056480

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056480

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4887
Formulation PBS containing 0.2% gelatin
Gene Accession No. NP_000901
Concentration 0.5mg/mL
Test Specificity Reacts with Human and Rhesus Monkey. 83% sequence identity with canine and mouse proteins.
Format Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mg
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Primate
Immunogen A portion of amino acids 350-400 of human NPY2-R was used as the immunogen.
Antigen NPY2R
Host Species Rabbit