Please login to your online account to display your discounted pricing

anti-NPM1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 21-53 of human Nucleophosmin were used as the immunogen.
Antigen NPM1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227157

 
novus biologicals canada
NBP227157
Unlabeled 0.1mg Each for $255.80

NBP227157SS

 
novus biologicals canada
NBP227157SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Within the range of amino acids 21-53 of human Nucleophosmin were used as the immunogen.
Antigen NPM1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in many species including human, mouse and rat.
Gene ID (Entrez) 4869
Formulation 100μg in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL