Please login to your online account to display your discounted pricing

anti-NPM1, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human mutant Nucleophosmin (within residues 250-294). [Accession No. AAW67752]
Antigen NPM1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11061646

 
novus biologicals canada
NB11061646
Unlabeled 0.1mL Each for $329.00

NB11061646T

 
novus biologicals canada
NB11061646SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal region of human mutant Nucleophosmin (within residues 250-294). [Accession No. AAW67752]
Antigen NPM1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This antibody is specific for the mutant form of Nucleophosmin.
Gene ID (Entrez) 4869
Formulation 100mM glycine, 65mM Tris, pH 7.0 in 50% glycerol
Concentration 0.3mg/mL