Please login to your online account to display your discounted pricing

NPC1-Like 1 Polyclonal Antibody, Invitrogen™


NPC1-Like 1 Antibody, PA1-16801, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Immunoprecipitation, Western Blot.

Manufacturer: pierce biotechnology inc  PA116801

Catalog No. APPA6801

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q9UHC9, Q6T3U4, Q6T3U3
Formulation 100μL purified antibody (1mg/mL) in tris citrate and 0.01% sodium azide
Gene ID (Entrez) 29881, 237636, 432367
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blot
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG
Storage Requirements Store at 4° C
Species Reactivity Human, Mouse, Rat
Gene Alias NPC3, Niemann-Pick C3, NPC1L1, Niemann-Pick type C1 Like 1 Protein, BC 400-127
Gene Symbols NPC1L1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of rat NPC1L1 (between residues 500-600).
Format Purified
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Quantity 100μL
Antigen NPC1-Like 1

NPC1-Like 1 Antibody, PA1-16801, from Invitrogen. Species Reactivity: Human, Mouse, Rat; Applications: Immunohistochemistry, Immunoprecipitation, Western Blot.

Dietary consumption and intestinal cholesterol absorption contribute to plasma cholesterol levels, which factor into the risk of coronary heart disease. Niemann-Pick type C1 Like 1 protein (NPC1L1 or NPC3) is enriched in the small intestine and is found in the brush border membrane of enterocytes. It plays a critical role in absorption of intestinal cholesterol.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: A synthetic peptide made to an internal region of rat NPC1L1 (between residues 500-600).
Formulation: 100μL purified antibody (1mg/mL) in tris citrate and 0.01% sodium azide
Synonyms: NPC3, Niemann-Pick C3, NPC1L1, Niemann-Pick type C1 Like 1 Protein, BC 400-127
Uniprot Accession: Q9UHC9, Q6T3U4, Q6T3U3
Entrez GeneID: 29881, 237636, 432367
Gene Symbol: NPC1L1