Please login to your online account to display your discounted pricing

anti-NOXA Polyclonal, Thermo Scientific™

NOXA Polyclonal antibody PA1-18331 from Thermo Scientific™ specifically detects NOXA in Mouse samples, and it is validated for Western Blot

$506.05 - $543.75

Specifications

Antigen NOXA
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-585

 
pierce biotechnology inc
PA118332
Unlabeled 100μL Each for $506.05

01-670-584

 
pierce biotechnology inc
PA118331
Unlabeled 100μL Each for $543.75
Description & Specifications

Specifications

Antigen NOXA
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C(75) A Q L R R I G D K V N L R Q K(90) of mouse Noxa.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

NOXA Polyclonal antibody PA1-18331 from Thermo Scientific™ specifically detects NOXA in Mouse samples, and it is validated for Western Blot

The Bcl-2 family of proteins which regulate apoptosis share identical sequences called Bcl-2 Homology domains (BH1-4). The BH3 proteins, including BID, NOXA, PUMA, BIK, BIM and BAD are all pro-apoptotic and share sequence identity within the amphipathic alpha-helical BH3 region, which is essential for their apoptotic function. NOXA is highly expressed in adult T-cell leukemia cell line.
Host Species: Rabbit
Cross Reactivity: Mouse
Isotype: IgG
Immunogen: Synthetic peptide corresponding to the N-terminal residues M(1) P G R K A R R N A P V N P T R(16) of mouse Noxa.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: PMAIP1, phorbol-12-myristate-13-acetate-induced protein 1, adult T cell leukemia-derived PMA-responsive, Immediate-early-response protein APR, PMA-ind
Uniprot Accession: Q9JM54
Gene Symbol: Pmaip1
Entrez GeneID: 58801
PA1-18331 detects NOXA in Mouse samples. PA1-18331 has been successfully used in Western Blot procedures. PA1-18331 The immunogen is Synthetic peptide corresponding to the N-terminal residues M(1) P G R K A R R N A P V N P T R(16) of mouse Noxa..