Please login to your online account to display your discounted pricing

anti-Notch-4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10093551SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10093551T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q99466
Gene ID (Entrez) 4855
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Rabbit
Antigen Notch-4
Format Affinity Purified
Test Specificity Notch 4 homolog.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 1mg/mL
Formulation PBS
Conjugate Unlabeled
Immunogen A synthetic peptide made to a C-terminal portion of human Notch4 (between amino acids 1900-2003) [UniProt Q99466]
Cross Reactivity Human
Regulatory Status RUO
Quantity 0.025mg