Please login to your online account to display your discounted pricing

anti-Notch-4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10093551

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10093551

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Host Species Rabbit
Formulation PBS
Primary or Secondary Primary
Test Specificity Notch 4 homolog.
Gene Accession No. Q99466
Concentration 1mg/mL
Regulatory Status RUO
Gene ID (Entrez) 4855
Conjugate Unlabeled
Antigen Notch-4
Quantity 0.1mg
Immunogen A synthetic peptide made to a C-terminal portion of human Notch4 (between amino acids 1900-2003) [UniProt Q99466]