Please login to your online account to display your discounted pricing

anti-Notch-1, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Within the range of amino acids 1755-1780 of human Notch1 were used as the immunogen.
Antigen Notch-1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224771

 
novus biologicals canada
NBP224771
Unlabeled 0.1mg Each for $307.00

NBP224771SS

 
novus biologicals canada
NBP224771SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Within the range of amino acids 1755-1780 of human Notch1 were used as the immunogen.
Antigen Notch-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human.
Gene Accession No. P46531
Gene ID (Entrez) 4851
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL