Please login to your online account to display your discounted pricing

anti-Nogo B receptor, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Isotype IgG
Immunogen 2006). This immunogen sequence is within the 93 amino acid putative ectodomain of the NgBR.
Antigen Nogo B receptor
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224950SS

 
novus biologicals canada
NBP224950SS
Unlabeled 0.025mg Each for $139.00

NBP224950

 
novus biologicals canada
NBP224950
Unlabeled 0.1mg Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen 2006). This immunogen sequence is within the 93 amino acid putative ectodomain of the NgBR.
Antigen Nogo B receptor
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene ID (Entrez) 116150
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL