Please login to your online account to display your discounted pricing

anti-NOD2, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human NOD2 (between residues 1000-1040). [UniProt No. Q9HC29]
Antigen NOD2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500253

 
novus biologicals canada
NB500253
Unlabeled 0.1mL Each for $329.00

NB500253SS

 
novus biologicals canada
NB500253SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the C-terminal region of human NOD2 (between residues 1000-1040). [UniProt No. Q9HC29]
Antigen NOD2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blot
Test Specificity Human. Predicted to react with monkey based on 100% sequence homology.
Gene Accession No. Q9HC29
Gene ID (Entrez) 64127
Formulation Tris-glycine, 150mM NaCl
Concentration 0.87mg/mL