Please login to your online account to display your discounted pricing

anti-NOD2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500253SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500253SS

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human. Predicted to react with monkey based on 100% sequence homology.
Formulation Tris-glycine, 150mM NaCl
Concentration 0.87mg/mL
Conjugate Unlabeled
Host Species Rabbit
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Regulatory Status RUO
Gene ID (Entrez) 64127
Cross Reactivity Human
Gene Accession No. Q9HC29
Monoclonal or Polyclonal Polyclonal
Antigen NOD2
Primary or Secondary Primary
Quantity 0.025mL
Format Affinity Purified
Immunogen A synthetic peptide made to the C-terminal region of human NOD2 (between residues 1000-1040). [UniProt No. Q9HC29]