missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-NOD2, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 125-162 of human Nod2.
Antigen NOD2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP227118 Novus Biologicals Canada
NBP227118
0.1mL Each for $259.02
NBP227118SS Novus Biologicals Canada
NBP227118SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
NOD2
Antisera
RUO
Researchers are encouraged to optimize dilutions for their specific testing, and to use the sequence information and accession number (NP_071445.1) that was used to design the peptide immunogen in order to assess potential cross-reactivity of the antibody. The amino acid sequence used as immunogen is 100% homologous in human, orangutan and chimp, 93% homologous in rhesus monkey, pig, cow and dog, and 86% homologous in mouse and rat.
64127
This antibody was developed against a synthetic peptide corresponding to amino acids 125-162 of human Nod2.
Polyclonal
Primary
Immunoprecipitation,Western Blot
Q9HC29
0.1ml; rabbit sera in glycerol.