Please login to your online account to display your discounted pricing

anti-NOD2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500253

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500253

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Western Blotting
Gene ID (Entrez) 64127
Regulatory Status RUO
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Test Specificity Human. Predicted to react with monkey based on 100% sequence homology.
Concentration 0.87mg/mL
Gene Accession No. Q9HC29
Immunogen A synthetic peptide made to the C-terminal region of human NOD2 (between residues 1000-1040). [UniProt No. Q9HC29]
Format Affinity Purified
Cross Reactivity Human
Quantity 0.1mL
Antigen NOD2
Host Species Rabbit